missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROR1 Mouse anti-Human, CoraLite 488, Clone: 1F5D8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66923-100UL
This item is not returnable.
View return policy
Description
ROR1 Monoclonal antibody apecifically detetects ROR1 antigen in Human Samples. It is validated for Flow Cytometry.Specifications
| ROR1 | |
| Monoclonal | |
| 1000 μg/mL | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| ROR1 | |
| ROR1 fusion protein Ag14586 | |
| 100 μL | |
| 4919 | |
| Store at -20°C. Avoid exposure to light. | |
| IgG1 |
| Flow Cytometry | |
| 1F5D8 | |
| CoraLite 488 | |
| BC006374 | |
| Mouse | |
| Protein G purification | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction