missing translation for 'onlineSavingsMsg'
Learn More

ROR1 Mouse anti-Human, CoraLite 488, Clone: 1F5D8, Proteintech

Mouse Monoclonal Antibody

£128.70 - £319.04

Specifications

Antigen ROR1
Clone 1F5D8
Applications Flow Cytometry
Classification Monoclonal
Conjugate CoraLite 488
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17751299
View Documents
Proteintech
CL488-66923-20UL
20 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
17619876
View Documents
Proteintech
CL488-66923-100UL
100 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

ROR1 Monoclonal antibody apecifically detetects ROR1 antigen in Human Samples. It is validated for Flow Cytometry.
Specifications

Specifications

ROR1
Flow Cytometry
CoraLite 488
Mouse
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
4919
ROR1 fusion protein Ag14586
Primary
Store at -20°C. Avoid exposure to light.
1F5D8
Monoclonal
Liquid
Human
BC006374
ROR1
IgG1
Protein G purification
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.