missing translation for 'onlineSavingsMsg'
Learn More

Ropinirole N-Oxide, TRC

Product Code. 30138086
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
10 mg
Unit Size:
10mg
1mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138086 10 mg 10mg
30135135 2.5 mg 2.5mg
17998324 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138086 Supplier Toronto Research Chemicals Supplier No. TRCR44101510MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1076199-41-1
Molecular Formula C16 H24 N2 O2
Molecular Weight (g/mol) 276.37
Synonym 2H-Indol-2-one, 1,3-dihydro-4-[2-(oxidodipropylamino)ethyl]-, Ropinirole N-Oxide
IUPAC Name N-[2-(2-oxo-1,3-dihydroindol-4-yl)ethyl]-N-propylpropan-1-amine oxide
SMILES CCC[N+]([O-])(CCC)CCc1cccc2NC(=O)Cc12

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C16H24N2O2/c1-3-9-18(20,10-4-2)11-8-13-6-5-7-15-14(13)12-16(19)17-15/h5-7H,3-4,8-12H2,1-2H3,(H,17,19)
Formula Weight 276.1838
Percent Purity >95%
Chemical Name or Material Ropinirole N-Oxide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.