CAS RN 1076199-41-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ropinirole N-Oxide, TRC
CAS: 1076199-41-1 Molecular Formula: C16 H24 N2 O2 Molecular Weight (g/mol): 276.37 Synonym: 2H-Indol-2-one, 1,3-dihydro-4-[2-(oxidodipropylamino)ethyl]-,Ropinirole N-Oxide IUPAC Name: N-[2-(2-oxo-1,3-dihydroindol-4-yl)ethyl]-N-propylpropan-1-amine oxide SMILES: CCC[N+]([O-])(CCC)CCc1cccc2NC(=O)Cc12