missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROCK1 Recombinant Rabbit Monoclonal Antibody (23GB2065), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | ROCK1 |
|---|---|
| Clone | 23GB2065 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088674
![]() |
Invitrogen
MA552474 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087683
|
Invitrogen
MA552475 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ROCK1 Recombinant Monoclonal antibody specifically detects ROCK1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| ROCK1 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P70335, Q13464, Q63644 | |
| 19877, 6093, 81762 | |
| IgG | |
| Affinity Chromatography | |
| Antibody |
| 23GB2065 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 1110055K06Rik; Ac2-154; Liver regeneration-related protein LRRG199; MGC131603; MGC43611; p150 RhoA-binding kinase ROK beta; p160 ROCK-1; p160ROCK; p160-ROCK; PRO0435; Renal carcinoma antigen NY-REN-35; Rho associated coiled-coil containing protein kinase 1; Rho kinase; Rho-associated coiled-coil containing protein kinase 1; Rho-associated coiled-coil forming kinase 1; Rho-associated kinase beta; rho-associated protein kinase 1; Rho-associated, coiled-coil containing protein kinase 1; rho-associated, coiled-coil-containing protein kinase 1; Rho-associated, coiled-coil-containing protein kinase I; ROCK1; ROCK-I | |
| A synthesized peptide derived from human ROCK1 (1100-1200AA). | |
| Primary | |
| -20°C | |
| Rock1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title