missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROCK1 Recombinant Rabbit Monoclonal Antibody (23GB2065), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen MA552475
This item is not returnable.
View return policy
Description
ROCK1 Recombinant Monoclonal antibody specifically detects ROCK1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot
Specifications
| ROCK1 | |
| Recombinant Monoclonal | |
| 0.8 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P70335, Q13464, Q63644 | |
| Rock1 | |
| A synthesized peptide derived from human ROCK1 (1100-1200AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB2065 | |
| Unconjugated | |
| Rock1 | |
| 1110055K06Rik; Ac2-154; Liver regeneration-related protein LRRG199; MGC131603; MGC43611; p150 RhoA-binding kinase ROK beta; p160 ROCK-1; p160ROCK; p160-ROCK; PRO0435; Renal carcinoma antigen NY-REN-35; Rho associated coiled-coil containing protein kinase 1; Rho kinase; Rho-associated coiled-coil containing protein kinase 1; Rho-associated coiled-coil forming kinase 1; Rho-associated kinase beta; rho-associated protein kinase 1; Rho-associated, coiled-coil containing protein kinase 1; rho-associated, coiled-coil-containing protein kinase 1; Rho-associated, coiled-coil-containing protein kinase I; ROCK1; ROCK-I | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 19877, 6093, 81762 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction