missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROCK1 Antibody (1H4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£161.00 - £435.00
Specifications
| Antigen | ROCK1 |
|---|---|
| Clone | 1H4 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000 |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18184829
|
Novus Biologicals
NBP2-37533 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18680776
|
Novus Biologicals
NBP2-37533-0.025ml |
0.025 mL |
£161.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ROCK1 Monoclonal specifically detects ROCK1 in Human samples. It is validated for Western Blot, ELISA.Specifications
| ROCK1 | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| Human | |
| EC 2.7.11, EC 2.7.11.1, MGC131603, MGC43611, p160 ROCK-1, p160ROCK, p160-ROCK, PRO0435, Renal carcinoma antigen NY-REN-35, Rho kinase, rho-associated protein kinase 1, Rho-associated, coiled-coil containing protein kinase 1, Rho-associated, coiled-coil-containing protein kinase 1 | |
| ROCK1 | |
| IgG1 | |
| Affinity Purified |
| 1H4 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
| Monoclonal | |
| Mouse | |
| Apoptosis, Breast Cancer, Cancer, Signal Transduction, Wnt Signaling Pathway | |
| PBS with 0.05% Sodium Azide | |
| 6093 | |
| Purified recombinant fragment of human ROCK1 expressed in E. Coli. | |
| Primary | |
| 158 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title