missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROCK1 Antibody (1H4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37533
This item is not returnable.
View return policy
Description
ROCK1 Monoclonal specifically detects ROCK1 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| ROCK1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
| EC 2.7.11, EC 2.7.11.1, MGC131603, MGC43611, p160 ROCK-1, p160ROCK, p160-ROCK, PRO0435, Renal carcinoma antigen NY-REN-35, Rho kinase, rho-associated protein kinase 1, Rho-associated, coiled-coil containing protein kinase 1, Rho-associated, coiled-coil-containing protein kinase 1 | |
| Mouse | |
| 158 kDa | |
| 0.1 mL | |
| Apoptosis, Breast Cancer, Cancer, Signal Transduction, Wnt Signaling Pathway | |
| 6093 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 1H4 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| ROCK1 | |
| Purified recombinant fragment of human ROCK1 expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction