Learn More
BRAND™ Robotic Tips, BIO-CERT™ Liquid Handling, Sterile
Product Code. 17586696 Shop All Brand Products
Change view
Click to view available options
Volume (Metric):
1 to 50 μL
10 to 200 μL
10 to 300 μL
40 to 1000 μL
Unit Size:
Pack of 960
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
17586696 40 to 1000 μL Pack of 960
17506696 1 to 50 μL Pack of 960
17566696 10 to 200 μL Pack of 960
17576696 10 to 300 μL Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17586696 Supplier BRAND™ Supplier No. 734172
  • ONLINE EXCLUSIVE PRICE
    £87.80
    £86.25 / Pack of 960
    Save £1.55
    1% Off
View all promotions
Estimated Shipment: 04-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Pipette tips offer reliable quality for precise and demanding analyses with the pipetting robot Liquid Handling Station.
  • The raw materials are free from DiHEMDA und Oleamid. Tips are manufactured at BRAND under monitored cleanroom conditions and are automatically packaged. No parting agents or demolding aids are used. All racked tips and filter tips up to 1000 μL are free of DNA (< 40 fg), RNase (< 8.6 fg), endotoxins (< 1 pg) and ATP (< 1 fg).
  • For use with pipetting robots, tips are required that are matched to the robot's mode of operation in terms of geometry and surface properties. BRAND robotic pipette tips and robotic filter tips meet all requirements for use with the Liquid Handling Station pipetting robot.
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 40 to 1000 μL
Filtered Filtered
Sterility Sterile
Packaging TipRack (960/Pk.)
Product Type Robotic Tips
Material Polypropylene