missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Robotic Sterile Pipette Tips

Product Code. 17294974 Shop All SP Scienceware Products
Click to view available options
For Use With (Application):
For: Hamilton, Volume: 100 μL, Conductive, Material: carbon filled polypropylene
For: Hamilton, Volume: 100 μL, Non-conductive, Material: Clear polypropylene
For: Hamilton, Volume: 50 μL, Conductive, Material: carbon filled polypropylene
For: Hamilton, Volume: 50 μL, Non-conductive, Material: clear polypropylene
For: Tecan, Volume: 100 μL, Conductive, Material: carbon filled polypropylene
For: Tecan, Volume: 100 μL, Non-conductive, Material: clear polypropylene
For: Tecan, Volume: 50 μL, Conductive, Material: carbon filled polypropylene
For: Tecan, Volume: 50 μL, Non-conductive, Material: clear polypropylene
Unit Size:
Pack of 16
Pack of 24
This item is not returnable. View return policy

Product Code. 17294974

Brand: Bel-Art™ F378601100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Designed for use with automated liquid handling equipment

  • Sterile, disposable, filtered pipette tips can be used in robotic systems
    • Filtered to prevent contamination of pipette arm
  • Conductive tip models detect liquid level when pipetting conductive liquids
    • Ensures repeatable volume accuracy
  • DNase, RNase, pyrogen free, manufactured in Class 8 clean room
  • CE marked

Specifications

Volume (Metric) 1000 μL
Filtered Filtered
Sterility Sterile
Color Clear
Autoclavable Not Autoclavable
Certifications/Compliance CE marked
For Use With (Application) For: Hamilton, Volume: 100 μL, Non-conductive, Material: Clear polypropylene
Format Box
Tip Style Standard
Packaging 16 racks x 96 tips
Quantity 1536
Product Type Robotic pipette tips
Conductivity No
Disposable Single-use
For Use With (Equipment) Hamilton STAR, STARlet and NIMBUS
Graduated No
Material Polypropylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.