missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies RoboSep™ Mouse Plasmacytoid DC Isolation Kit
Click to view available options
Quantity:
2 x 109 Cells
Unit Size:
1 set
Description
- The EasySep™ Mouse Plasmacytoid DC Isolation Kit is designed to isolate plasmacytoid dendritic cells (pDCs) from single-cell suspensions of splenocytes or other tissues by negative selection
- Unwanted cells are targeted for removal with biotinylated antibodies that are directed against non-pDCs
- Labeled cells are then recognized by Tetrameric Antibody Complexes that are directed against biotin and dextran
- These cells are bound to magnetic particles and separated using an EasySep™ magnet without the use of columns
- Desired cells are poured off into a new tube
Specifications
Specifications
| Cell Type | Dendritic Cells |
| For Use With (Application) | Cell Isolation |
| Quantity | 2 x 109 Cells |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction