missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies RoboSep™ Mouse NK Cell Isolation Kit

Product Code. 17107981
Encompass
Click to view available options
Quantity:
1 x 109 cells
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 17107981

Brand: STEMCELL Technologies 19855RF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

EasySep™ kit with RoboSep™ buffer and RoboSep™ filter tips

  • The EasySep™ Mouse NK Cell Isolation Kit targets non-NK cells by labeling unwanted cells with biotinylated antibodies and EasySep™ Streptavidin RapidSpheres™, and separates cells without columns using an EasySep™ magnet
  • Desired cells are simply poured off into a new tube
  • Isolated cells are immediately available for downstream applications such as flow cytometry, culture or cell-based assays
  • This product replaces the EasySep™ Mouse NK Cell Enrichment Kit (Catalog #19755) for even faster cell isolations

Specifications

Cell Type NK Cells
For Use With (Application) Cell Isolation
Quantity 1 x 109 cells
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.