missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies RoboSep™ Mouse CD4+CD62L+ T Cell Isolation Kit
Click to view available options
Quantity:
1 x 109 cells
Unit Size:
1 set
Description
- Isolate highly purified naïve CD4+ T cells from single-cell suspensions of splenocytes or other tissues by using a simple, two-step method
- First, naive CD4+ T cells are pre-enriched by negative selection using the EasySep™ Mouse Naïve CD4+ T Cell Pre-Enrichment Cocktail containing biotinylated antibodies directed against non-naïve CD4+ T cells (CD8, CD11b, CD11c, CD19, CD24, CD25, CD45R, CD49b, TCRγ/δ, TER119)
- Then CD4+CD62L+ cells, pre-labeled with CD62L PE, are selected using the EasySep™ PE Selection Cocktail
- The desired cells are labeled with antibodies and magnetic particles
- The cells are separated without columns using an EasySep™ magnet
- For the isolation of mouse naïve CD4+ T cells in as little as 15 minutes by negative selection, we recommend using the EasySep™ Mouse Naïve CD4+ T Cell Isolation Kit (19765)
Specifications
Specifications
| Cell Type | T Cells, T Cells, CD4+ |
| For Use With (Application) | Cell Isolation |
| Quantity | 1 x 109 cells |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction