Learn More
STEMCELL Technologies RoboSep™ Human Myeloid Positive Selection Kit II
Click to view available options
Quantity:
1 x 109 Cells
Unit Size:
1 set
Description
- The EasySep™ Human Myeloid Positive Selection Kit II is designed to isolate CD33+ and CD66b+ cells from ammonium chloride lysed blood or bone marrow by positive selection
- Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD33, CD66b and dextran-coated magnetic particles
- Labeled cells are separated using an EasySep™ magnet without the use of columns
- Cells of interest remain in the tube while unwanted cells are poured off
- The CD33 and CD66b antigens are expressed on myeloid progenitors monocytes and granulocytes
- This product replaces the EasySep™ Human Myeloid Positive Selection Kit (Catalog #18653) for even faster cell isolations
Specifications
Specifications
| Cell Type | Granulocytes and Subsets, Myeloid Cells |
| Quantity | 1 x 109 Cells |