missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNH1 Mouse anti-Human, Clone: 4A1C4, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | RNH1 |
|---|---|
| Clone | 4A1C4 |
| Concentration | 1.18 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16857535
|
Proteintech
66028-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16847535
|
Proteintech
66028-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RNH1 is the inhibitor of pancreatic RNase and angiogenin. RNH1 may also function in the modulation of cellular activities.Specifications
| RNH1 | |
| 1.18 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Placental RNase inhibitor, PRI, RAI, Ribonuclease inhibitor, RNH, RNH1 | |
| RNH1 | |
| IgG2b | |
| Protein A | |
| Antibody |
| 4A1C4 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P13489 | |
| 6050 | |
| RNH1 Fusion Protein Ag18093 | |
| Primary | |
| -20°C | |
| RNH1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title