missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNH1 Mouse anti-Human, Clone: 4A1C4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66028-1-IG-150UL
This item is not returnable.
View return policy
Description
RNH1 is the inhibitor of pancreatic RNase and angiogenin. RNH1 may also function in the modulation of cellular activities.Specifications
| RNH1 | |
| Monoclonal | |
| 1.18 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P13489 | |
| RNH1 | |
| RNH1 Fusion Protein Ag18093 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 4A1C4 | |
| Unconjugated | |
| RNH1 | |
| Placental RNase inhibitor, PRI, RAI, Ribonuclease inhibitor, RNH, RNH1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 6050 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction