missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF90 Monoclonal Antibody (K56013_18G5), Invitrogen™
Mouse Monoclonal Antibody
£380.00 - £570.00
Specifications
| Antigen | RNF90 |
|---|---|
| Clone | K56013_18G5 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30213009
|
Invitrogen™
MA555738 |
50 μL |
£380.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30203389
|
Invitrogen™
MA556118 |
200 μL |
£570.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: GQHGEPFKLY CQDDGRAICV VCDRAREHRE HAVLPLDEAV QEAKELLESR LRVLKKELED CEVFRSTEKK ESKELLKQMA AEQEKVGAEF QALRAFLVEQ EGRLLGRLEE LSREVAQKQN ENLAQLGVEI TQLSKLSSQI QETAQKPDLD
RNF90 Monoclonal antibody specifically detects RNF90 in Human samples. It is validated for Western BlotSpecifications
| RNF90 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| AI790312; E3 ubiquitin-protein ligase TRIM7; Glycogenin-interacting protein; GNIP; RING finger protein 90; RNF90; TRIM7; tripartite motif containing 7; tripartite motif protein 7; tripartite motif protein TRIM7; tripartite motif-containing 7; tripartite motif-containing protein 7 | |
| Recombinant human TRIM7 (immunogen range is 131-280). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Trim7 |
| K56013_18G5 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9C029 | |
| 81786 | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title