missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF90 Monoclonal Antibody (K56013_18G5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA556118
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: GQHGEPFKLY CQDDGRAICV VCDRAREHRE HAVLPLDEAV QEAKELLESR LRVLKKELED CEVFRSTEKK ESKELLKQMA AEQEKVGAEF QALRAFLVEQ EGRLLGRLEE LSREVAQKQN ENLAQLGVEI TQLSKLSSQI QETAQKPDLD
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1, a B-box type 2, and a coiled-coil region. The protein localizes to both the nucleus and the cytoplasm, and may represent a participant in the initiation of glycogen synthesis. Multiple transcript variants have been found for this gene, and some of them encode the same isoform.
Specifications
| RNF90 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q9C029 | |
| Trim7 | |
| Recombinant human TRIM7 (immunogen range is 131-280). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| K56013_18G5 | |
| Unconjugated | |
| Trim7 | |
| AI790312; E3 ubiquitin-protein ligase TRIM7; Glycogenin-interacting protein; GNIP; RING finger protein 90; RNF90; TRIM7; tripartite motif containing 7; tripartite motif protein 7; tripartite motif protein TRIM7; tripartite motif-containing 7; tripartite motif-containing protein 7 | |
| Mouse | |
| Protein A | |
| RUO | |
| 81786 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction