missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF17 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£155.00 - £364.00
Specifications
| Antigen | RNF17 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
RNF17 Polyclonal antibody specifically detects RNF17 in Human samples. It is validated for Western BlotSpecifications
| RNF17 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| FLJ11045, Mmip-2, ring finger protein 17, SPATA23, spermatogenesis associated 23, TDRD4, tudor domain containing 4, Tudor domain-containing protein 4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 900-1200 of human RNF17 (NP_112567.2). VSAKSLPNENFQSLYNKELPVHICNVISPEKIYVQWLLTENLLNSLEEKMIAAYENSKWEPVKWENDMHCAVKIQDKNQWRRGQIIRMVTDTLVEVLLYDVGVELVVNVDCLRKLEENLKTMGRLSLECSLVDIRPAGGSDKWTATACDCLSLYLTGAVATIILQVDSEENNTTWPLPVKIFCRDEKGERVDVSKYLIKKGLALRERRINNLDNSHSLSEKSLEVPLEQEDSVVTNCIKTNFDPDKKTADIISEQKVSEFQEKILEPRTTRGYKPPAIPNMNVFEATVSCVGDDGTIFVVP | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 56163 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title