missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF17 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94677-0.1ml
This item is not returnable.
View return policy
Description
RNF17 Polyclonal antibody specifically detects RNF17 in Human samples. It is validated for Western Blot
Specifications
| RNF17 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| FLJ11045, Mmip-2, ring finger protein 17, SPATA23, spermatogenesis associated 23, TDRD4, tudor domain containing 4, Tudor domain-containing protein 4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 900-1200 of human RNF17 (NP_112567.2). VSAKSLPNENFQSLYNKELPVHICNVISPEKIYVQWLLTENLLNSLEEKMIAAYENSKWEPVKWENDMHCAVKIQDKNQWRRGQIIRMVTDTLVEVLLYDVGVELVVNVDCLRKLEENLKTMGRLSLECSLVDIRPAGGSDKWTATACDCLSLYLTGAVATIILQVDSEENNTTWPLPVKIFCRDEKGERVDVSKYLIKKGLALRERRINNLDNSHSLSEKSLEVPLEQEDSVVTNCIKTNFDPDKKTADIISEQKVSEFQEKILEPRTTRGYKPPAIPNMNVFEATVSCVGDDGTIFVVP | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 56163 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction