missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF135 Monoclonal Antibody (K49037_14A5), Invitrogen™
Mouse Monoclonal Antibody
£380.00 - £570.00
Specifications
| Antigen | RNF135 |
|---|---|
| Clone | K49037_14A5 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30194043
|
Invitrogen™
MA556146 |
200 μL |
£570.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30202428
|
Invitrogen™
MA555766 |
50 μL |
£380.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: HLRKNTLLQD LADKYRRAAR EIQAGSDPAH CPCPGSSSLS SAAARPRRRP ELQRVAVEKS ITEVAQELTE LVEHLVDIVR SLQNQRPLSE SGPDNELSIL GKAFSSGVDL SMASPKLVTS DTAAGKIRDI LHDLEEIQEK LQESVTWKEA PEAQMQGELL
RNF135 Monoclonal antibody specifically detects RNF135 in Human samples. It is validated for Western BlotSpezifikation
| RNF135 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 0610037N03Rik; 2410006N06Rik; E3 ubiquitin-protein ligase RNF135; L13; LRRGT00184; MGC13061; MMFD; REUL; RIG-I E3 ubiquitin ligase; ring finger protein 135; RING finger protein 135; E3 ubiquitin-protein ligase RNF135; RING-type E3 ubiquitin transferase RNF135; Riplet; RNF135; U 2-3-0 | |
| Recombinant human RNF135 (immunogen range is 71-230). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| RNF135 |
| K49037_14A5 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q8IUD6 | |
| 84282 | |
| IgG1 | |
| Protein A | |
| Antibody |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts