missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNF135 Monoclonal Antibody (K49037_14A5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA556146
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: HLRKNTLLQD LADKYRRAAR EIQAGSDPAH CPCPGSSSLS SAAARPRRRP ELQRVAVEKS ITEVAQELTE LVEHLVDIVR SLQNQRPLSE SGPDNELSIL GKAFSSGVDL SMASPKLVTS DTAAGKIRDI LHDLEEIQEK LQESVTWKEA PEAQMQGELL
The protein encoded by RNF135 contains a RING finger domain, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions. This gene is located in a chromosomal region known to be frequently deleted in patients with neurofibromatosis.
Specifications
| RNF135 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q8IUD6 | |
| RNF135 | |
| Recombinant human RNF135 (immunogen range is 71-230). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| K49037_14A5 | |
| Unconjugated | |
| RNF135 | |
| 0610037N03Rik; 2410006N06Rik; E3 ubiquitin-protein ligase RNF135; L13; LRRGT00184; MGC13061; MMFD; REUL; RIG-I E3 ubiquitin ligase; ring finger protein 135; RING finger protein 135; E3 ubiquitin-protein ligase RNF135; RING-type E3 ubiquitin transferase RNF135; Riplet; RNF135; U 2-3-0 | |
| Mouse | |
| Protein A | |
| RUO | |
| 84282 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction