missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNase L Antibody (SR2243), Novus Biologicals™
Rabbit Monoclonal Antibody
£257.00 - £341.00
Specifications
| Antigen | RNase L |
|---|---|
| Clone | SR2243 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18640744
|
Novus Biologicals
NBP3-22080-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18610194
|
Novus Biologicals
NBP3-22080-50ul |
50 μg |
£257.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RNase L Monoclonal antibody specifically detects RNase L in Human samples. It is validated for Western BlotSpecifications
| RNase L | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 2-5A-dependent ribonuclease, 2-5A-dependent RNase, 2'-5'-oligoisoadenylate synthetase-dependent, EC 3.1.26.-, HPC1, interferon-induced 2-5A-dependent RNase, MGC104972, MGC133329, PRCA1, prostate cancer 1, Ribonuclease 4, Ribonuclease L, ribonuclease L (2'-5'-oligoisoadenylate synthetase-dependent), RNase L, RNS4DKFZp781D08126 | |
| A synthesized peptide derived from human RNase L (Uniprot #: Q05823) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR2243 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Chromatin Research, Cytokine Research, DNA replication Transcription Translation and Splicing, Immunology, Innate Immunity, Prostate Cancer, Protein Kinase | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 6041 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title