missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNase L Antibody (SR2243), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22080-50ul
219.30 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
RNase L Monoclonal antibody specifically detects RNase L in Human samples. It is validated for Western Blot
Specifications
| RNase L | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| SR2243 | |
| Western Blot 1:500-1:2000 | |
| 2-5A-dependent ribonuclease, 2-5A-dependent RNase, 2'-5'-oligoisoadenylate synthetase-dependent, EC 3.1.26.-, HPC1, interferon-induced 2-5A-dependent RNase, MGC104972, MGC133329, PRCA1, prostate cancer 1, Ribonuclease 4, Ribonuclease L, ribonuclease L (2'-5'-oligoisoadenylate synthetase-dependent), RNase L, RNS4DKFZp781D08126 | |
| A synthesized peptide derived from human RNase L (Uniprot #: Q05823) | |
| 50 μg | |
| Chromatin Research, Cytokine Research, DNA replication Transcription Translation and Splicing, Immunology, Innate Immunity, Prostate Cancer, Protein Kinase | |
| 6041 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction