missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNase L Antibody (2E9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 12 publications
£189.00 - £390.00
Specifications
| Antigen | RNase L |
|---|---|
| Clone | 2000000000 |
| Concentration | 1.0 mg/ml |
| Dilution | Western Blot 1:1000-1:4000 (ECL), ELISA 1:100-1:2000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, Knockout Validated |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoassay |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18080284
|
Novus Biologicals
NB100-351SS |
0.025 mL |
£189.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18765063
|
Novus Biologicals
NB100-351 |
0.1 mL |
£390.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RNase L Monoclonal specifically detects RNase L in Human, Hamster, Primate, Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| RNase L | |
| 1.0 mg/ml | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoassay | |
| Unconjugated | |
| Mouse | |
| Chromatin Research, Cytokine Research, DNA replication Transcription Translation and Splicing, Immunology, Innate Immunity, Prostate Cancer, Protein Kinase | |
| Q05823 | |
| 6041 | |
| Purified full-length recombinant human RNaseL produced in insect cells. [UniProt# Q05823] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| 83 kDa |
| 2000000000 | |
| Western Blot 1:1000-1:4000 (ECL), ELISA 1:100-1:2000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, Knockout Validated | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Hamster, Primate, Mouse (Negative), Rat (Negative) | |
| 2-5A-dependent ribonuclease, 2-5A-dependent RNase, 2'-5'-oligoisoadenylate synthetase-dependent, EC 3.1.26.-, HPC1, interferon-induced 2-5A-dependent RNase, MGC104972, MGC133329, PRCA1, prostate cancer 1, Ribonuclease 4, Ribonuclease L, ribonuclease L (2'-5'-oligoisoadenylate synthetase-dependent), RNase L, RNS4DKFZp781D08126 | |
| RNASEL | |
| IgG1 κ | |
| Protein G purified | |
| Human, hamster and primate. This antibody does not react with mouse or rat. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Produkttitel