missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNase L Antibody (2E9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 12 publications
Brand: Novus Biologicals NB100-351SS
This item is not returnable.
View return policy
Description
RNase L Monoclonal specifically detects RNase L in Human, Hamster, Primate, Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| RNase L | |
| Monoclonal | |
| 1.0 mg/ml | |
| Western Blot 1:1000-1:4000 (ECL), ELISA 1:100-1:2000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, Knockout Validated | |
| 2-5A-dependent ribonuclease, 2-5A-dependent RNase, 2'-5'-oligoisoadenylate synthetase-dependent, EC 3.1.26.-, HPC1, interferon-induced 2-5A-dependent RNase, MGC104972, MGC133329, PRCA1, prostate cancer 1, Ribonuclease 4, Ribonuclease L, ribonuclease L (2'-5'-oligoisoadenylate synthetase-dependent), RNase L, RNS4DKFZp781D08126 | |
| Mouse | |
| 83 kDa | |
| 0.025 mL | |
| Chromatin Research, Cytokine Research, DNA replication Transcription Translation and Splicing, Immunology, Innate Immunity, Prostate Cancer, Protein Kinase | |
| 6041 | |
| Human, Hamster, Primate, Mouse (Negative), Rat (Negative) | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoassay | |
| 2000000000 | |
| Unconjugated | |
| Q05823 | |
| RNASEL | |
| Purified full-length recombinant human RNaseL produced in insect cells. [UniProt# Q05823] | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, hamster and primate. This antibody does not react with mouse or rat. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction