missing translation for 'onlineSavingsMsg'
Learn More

RNA Helicase A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-56889

 View more versions of this product

Product Code. 18636728

  • £592.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

RNA Helicase A Polyclonal antibody specifically detects RNA Helicase A in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RNA Helicase A
Polyclonal
Western Blot 0.04-0.4 μg/mL
DDX9ATP-dependent RNA helicase A, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 9, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 9 (RNA helicase A, nuclear DNAhelicase II; leukophysin), DEAH (Asp-Glu-Ala-His) box polypeptide 9, DEAH box protein 9, EC 3.6.1, EC 3.6.1.15, EC 3.6.4.13, FLJ17406, leukophysin, LKP, NDH II, NDH2, NDHII, Nuclear DNA helicase II, RHA
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence: TIYIKQLGRRIFAREHGSNKKLAAQSCALSLVRQLYHLGVVEAYSGLTKKKEGETVEPYKVNLSQDLEHQLQNIIQELNLEILPPPEDPSVPVALNIGKLAQFEPSQ
100 μg
Core ESC Like Genes, Stem Cell Markers
1660
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Western Blot
Unconjugated
PBS (pH 7.2) and 40% Glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.