missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals RITA, p53 activator III
Click to view available options
Quantity:
1 mg
10 mg
Description
A cell-permeable, p53-targeting, tricyclic thiophene derivative
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 10 mg |
Content And Storage | 2°C to 8°C |
For Use With (Application) | A cell-permeable, p53-targeting, tricyclic thiophene derivative |
Formula Weight | 292.40g/mol |
Sensitivity | Light Sensitive |
Product Type | Signal Transduction Reagent |
Form | Crystalline solid |
InChI Key | KZENBFUSKMWCJF-UHFFFAOYSA-N |
SMILES | OCC1=CC=C(S1)C1=CC=C(O1)C1=CC=C(CO)S1 |
IUPAC Name | (5-{5-[5-(hydroxymethyl)thiophen-2-yl]furan-2-yl}thiophen-2-yl)methanol |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'