missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rift Valley Fever Virus Nucleocapsid Mouse anti-Virus, Clone: IA4.1.D5.B8, Novus Biologicals™
Mouse Monoclonal Antibody
£437.00
Specifications
| Antigen | Rift Valley Fever Virus Nucleocapsid |
|---|---|
| Clone | IA4.1.D5.B8 |
| Concentration | 1 mg/mL |
| Dilution | ELISA |
| Applications | ELISA |
Description
Rift Valley Fever Virus Nucleocapsid Monoclonal antibody specifically detects Rift Valley Fever Virus Nucleocapsid in Virus samples. It is validated for ELISASpecifications
| Rift Valley Fever Virus Nucleocapsid | |
| 1 mg/mL | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Infections (Virus Bacteria and Parasites) | |
| Phosphate Buffered Saline, pH 7.4, Filter Sterile | |
| 9538291 | |
| IgG1 κ | |
| Protein G purified | |
| Antibody |
| IA4.1.D5.B8 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Virus | |
| Fragment, NP, nucleocapsid, Nucleocapsid protein, Nucleocapsid proteinNucleoprotein, Nucleoprotein, RVFV_sS_gp2 | |
| Rift Valley Fever Virus Nucleoprotein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title