missing translation for 'onlineSavingsMsg'
Learn More

Rift Valley Fever Virus Nucleocapsid Mouse anti-Virus, Clone: IA4.1.D5.B8, Novus Biologicals™

Product Code. 18339066 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18339066 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18339066 Supplier Novus Biologicals Supplier No. NBP3148600.1MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Rift Valley Fever Virus Nucleocapsid Monoclonal antibody specifically detects Rift Valley Fever Virus Nucleocapsid in Virus samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rift Valley Fever Virus Nucleocapsid
Applications ELISA
Classification Monoclonal
Clone IA4.1.D5.B8
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA
Formulation Phosphate Buffered Saline, pH 7.4, Filter Sterile
Gene Alias Fragment, NP, nucleocapsid, Nucleocapsid protein, Nucleocapsid proteinNucleoprotein, Nucleoprotein, RVFV_sS_gp2
Host Species Mouse
Immunogen Rift Valley Fever Virus Nucleoprotein
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Gene ID (Entrez) 9538291
Target Species Virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.