missing translation for 'onlineSavingsMsg'
Learn More

Rifampicin, Fisher BioReagents™

Product Code. 10396743 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
Packaging:
Amber Glass
Unit Size:
1g
250mg
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10396743 5 g Amber Glass 5g
10103443 250 mg Amber Glass 250mg
10533325 1 g Amber Glass 1g
10562975 25 g Amber Glass 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10396743 Supplier Fisher BioReagents Supplier No. BP26795
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rifampicin

  • An antibiotic used to inhibit synthesis of proteins during recombinant protein expression in bacteria
  • Rif inhibits bacterial DNA-dependent RNA polymerase 
  • Rif can also be used as a selective agent for some strains
  • Mammalian RNA polymerase is not affected
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13292-46-1
Molecular Formula C43H58N4O12
Molecular Weight (g/mol) 822.953
InChI Key FZYOVNIOYYPUPY-HRJPTAQKSA-N
Synonym Rif
PubChem CID 131839595
SMILES CC1C=CC=C(C(=O)NC2=C(C3=C(C(=C4C(=C3C(=O)C2=CNN5CCN(CC5)C)C(=O)C(O4)(OC=CC(C(C(C(C(C(C1O)C)O)C)OC(=O)C)C)OC)C)C)O)O)C

Specifications

Type Rifampicin
Physical Form Solid
Melting Point 183°C
Loss on Drying < 1.0%
Packaging Amber Glass
Color Orange
Quantity 5 g
Assay Percent Range 97%
Merck Index 15, 8341
Formula Weight 822.96
Chemical Name or Material Rifampicin
Show More Show Less
compliance-icons
Product Identifier
  • Rifampin
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.