missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ribosomal Protein L17 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-74075
This item is not returnable.
View return policy
Description
Ribosomal Protein L17 Polyclonal specifically detects Ribosomal Protein L17 in Mouse samples. It is validated for Western Blot.
Specifications
| Ribosomal Protein L17 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| FLJ18762,60S ribosomal protein L23, FLJ92089, gene encoding putative NFkB activating protein, MGC117162, PD-1, ribosomal protein L17, rpL23, RPL23,60S ribosomal protein L17 | |
| Rabbit | |
| 20 kDa | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Bovine: 100%; Chicken: 100%; Canine: 100%; Goat: 100%; Mouse: 100%; Rabbit: 100%; Zebrafish: 100%; Xenopus: 92%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Goat, Rabbit, Sheep, Zebrafish | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9CPR4 | |
| RPL17 | |
| Synthetic peptides corresponding to the N terminal of Rpl17. Immunizing peptide sequence SLDPENPTKSCKSRGSNLRVHFKNTRETAQAIKGMHIRKATKYLKDVTLK. | |
| Affinity purified | |
| RUO | |
| 6139 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction