missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Rhodamine B

Product Code. 16469406
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16469406 100 mg 100mg
16463459 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16469406 Supplier MedChemExpress Supplier No. HYY0016
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Rhodamine B is a staining fluorescent dye, commonly used for dyeing textiles, paper, soap, leather, and drugs.

Specifications

Content And Storage 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light)
Quantity 100 mg
Shipping Condition Room temperature
Solubility DMSO : 125 mg/mL (260.95 mM; Need ultrasonic) ∣ H2O : 100 mg/mL (208.76 mM; Need ultrasonic)
Target NO Synthase ∣ MMP ∣ Endogenous Metabolite
Product Type Dye Reagents
Molecular Weight (g/mol) 479.01 g/mol
CAS 81-88-9
Synonym Basic Violet 10 ∣ Brilliant Pink B ∣ Rhodamine O ∣ Tetraethylrhodamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.