missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Rhodamine 6G

Product Code. 16482419
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16482419 1 g 1g
16461168 5 g 5g
16439895 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16482419 Supplier MedChemExpress Supplier No. HYD0309
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Rhodamine 6G is a rhodamine analog useful in Pgp efflux assays. It can be used in characterizing the kinetics of MRP1- mediated efflux, and as a laser dye and potential mitochondrial probe.

Specifications

Content And Storage 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light)
Quantity 1 g
Shipping Condition Room temperature
Solubility DMSO : 25 mg/mL (52.19 mM; Need ultrasonic) ∣ H2O : 10 mg/mL (20.88 mM; ultrasonic and warming and heat to 60°C)
Target HBV
Product Type Dye Reagents
Molecular Weight (g/mol) 479.01 g/mol
CAS 989-38-8
Synonym Basic Red 1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.