missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGS5 Antibody (CL5568), Novus Biologicals™
Mouse Monoclonal Antibody
£222.00 - £470.00
Specifications
| Antigen | RGS5 |
|---|---|
| Clone | CL5568 |
| Dilution | Western Blot 1:500 - 1:1000, Immunohistochemistry-Paraffin |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18243952
|
Novus Biologicals
NBP2-61419 |
100 μL |
£470.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18693058
|
Novus Biologicals
NBP2-61419-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RGS5 Monoclonal specifically detects RGS5 in Human samples. It is validated for Western Blot.Specifications
| RGS5 | |
| Western Blot 1:500 - 1:1000, Immunohistochemistry-Paraffin | |
| Unconjugated | |
| Mouse | |
| MST092, MST106, MST129, MSTP032, MSTP092, MSTP106, MSTP129, regulator of G-protein signaling 5, regulator of G-protein signalling 5 | |
| RGS5 | |
| IgG1 | |
| Protein A purified |
| CL5568 | |
| Monoclonal | |
| Purified | |
| Signal Transduction | |
| 8490 | |
| This antibody was developed against a synthetic peptide corresponding to amino acids: GDLVIPYNEKPEKPAKTQK | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title