missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGS5 Antibody (CL5568), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61419-25ul
This item is not returnable.
View return policy
Description
RGS5 Monoclonal specifically detects RGS5 in Human samples. It is validated for Western Blot.
Specifications
| RGS5 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.2), containing 40% glycerol with 0.02% Sodium Azide | |
| RGS5 | |
| This antibody was developed against a synthetic peptide corresponding to amino acids: GDLVIPYNEKPEKPAKTQK | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL5568 | |
| Western Blot 1:500 - 1:1000, Immunohistochemistry-Paraffin | |
| MST092, MST106, MST129, MSTP032, MSTP092, MSTP106, MSTP129, regulator of G-protein signaling 5, regulator of G-protein signalling 5 | |
| Mouse | |
| Protein A purified | |
| Signal Transduction | |
| 8490 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction