missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGS16 Antibody - BSA Free, Novus Biologicals™
Chicken Polyclonal Antibody has been used in 1 publication
£288.00
Specifications
| Antigen | RGS16 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:200-1:500, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
RGS16 Polyclonal specifically detects RGS16 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
| RGS16 | |
| Western Blot 1:200-1:500, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen | |
| Polyclonal | |
| Chicken | |
| Signal Transduction | |
| PBS with 0.02% Sodium Azide | |
| A28-RGS14, A28-RGS14P, hRGS-r, regulator of G-protein signaling 16, regulator of G-protein signalling 16, Retinally abundant regulator of G-protein signaling, Retinal-specific RGS, RGSR, RGS-R | |
| RGS16 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| O15492 | |
| 6004 | |
| Synthetic peptide: corresponding to amino acids 130-202 of Human RGS16 (NP_002919.1). | |
| Affinity Purified | |
| RGS16 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title