missing translation for 'onlineSavingsMsg'
Learn More

RGS16 Antibody - BSA Free, Novus Biologicals™

Chicken Polyclonal Antibody has been used in 1 publication

Brand:  Novus Biologicals NB300-350-0.025mg

 View more versions of this product

Product Code. 18527591

  • £288.00 / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

RGS16 Polyclonal specifically detects RGS16 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RGS16
Polyclonal
Unconjugated
PBS with 0.02% Sodium Azide
A28-RGS14, A28-RGS14P, hRGS-r, regulator of G-protein signaling 16, regulator of G-protein signalling 16, Retinally abundant regulator of G-protein signaling, Retinal-specific RGS, RGSR, RGS-R
Chicken
Affinity Purified
RUO
Primary
RGS16
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Western Blot
1 mg/mL
Western Blot 1:200-1:500, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen
O15492
RGS16
Synthetic peptide: corresponding to amino acids 130-202 of Human RGS16 (NP_002919.1).
0.025 mg
Signal Transduction
6004
Human
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only