missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGM-A Antibody (SR1626), Novus Biologicals™
Rabbit Monoclonal Antibody
£257.00 - £341.00
Specifications
| Antigen | RGM-A |
|---|---|
| Clone | SR1626 |
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18609144
|
Novus Biologicals
NBP3-22449-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18649244
|
Novus Biologicals
NBP3-22449-50ul |
50 μg |
£257.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RGM-A Monoclonal antibody specifically detects RGM-A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| RGM-A | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| RGM domain family member A, RGM domain family, member A, RGMrepulsive guidance molecule A | |
| A synthesized peptide derived from human RGM-A (Uniprot #: Q96B86) | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| SR1626 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Neuroscience, Stem Cells | |
| PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
| 56963 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title