missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGM-A Antibody (SR1626), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22449-50ul
This item is not returnable.
View return policy
Description
RGM-A Monoclonal antibody specifically detects RGM-A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| RGM-A | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunofluorescence | |
| SR1626 | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| RGM domain family member A, RGM domain family, member A, RGMrepulsive guidance molecule A | |
| A synthesized peptide derived from human RGM-A (Uniprot #: Q96B86) | |
| 50 μg | |
| Neuroscience, Stem Cells | |
| 56963 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction