missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RG9MTD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-57361
This item is not returnable.
View return policy
Description
RG9MTD1 Polyclonal specifically detects RG9MTD1 in Human samples. It is validated for Western Blot.
Specifications
| RG9MTD1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| EC 2.1.1, EC 2.1.1.-, EC 2.1.1.36, FLJ20432, HBV pre-S2 trans-regulated protein 2, Mitochondrial RNase P protein 1, MRPP1mitochondrial ribonuclease P protein 1, NY-REN-49 antigen, Renal carcinoma antigen NY-REN-49, RNA (guanine-9-) methyltransferase domain containing 1, RNA (guanine-9-)-methyltransferase domain-containing protein 1 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 54931 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q7L0Y3 | |
| TRMT10C | |
| Synthetic peptides corresponding to RG9MTD1 (RNA (guanine-9-) methyltransferase domain containing 1) The peptide sequence was selected from the middle region of RG9MTD1. Peptide sequence MKRKELQNTVSQLLESEGWNRRNVDPFHIYFCNLKIDGALHRELVKRYQE. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Human: 100%; Equine: 92%; Guinea pig: 91%; Pig: 85%; Rabbit: 85%; Zebrafish: 84%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction