missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFPL4B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-55032
This item is not returnable.
View return policy
Description
RFPL4B Polyclonal specifically detects RFPL4B in Human samples. It is validated for Western Blot.
Specifications
| RFPL4B | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| ret finger protein-like 4B, RNF211RING finger protein 211 | |
| Rabbit | |
| 30 kDa | |
| 100 μL | |
| Zinc Finger | |
| 442247 | |
| Human | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q6ZWI9 | |
| RFPL4B | |
| Synthetic peptides corresponding to RFPL4B(ret finger protein-like 4B) The peptide sequence was selected from the middle region of RFPL4B. Peptide sequence EVGEVKSWSLGVCKEPADRKSNDLFPEHGFWISMKAGAIHANTHLERIPA. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction