missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFP Rat, Unconjugated, Clone: 5F8, Proteintech ChromoTek
Rat Monoclonal Antibody
£137.40 - £178.00
Specifications
| Antigen | RFP |
|---|---|
| Clone | 5F8 |
| Applications | Immunofluorescence, ELISA |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
Rat monoclonal [5F8] to Red Fluorescent Proteins (RFP)
Specifications
| RFP | |
| Immunofluorescence, ELISA | |
| Unconjugated | |
| IgG2a | |
| Shipped at ambient temperature. Store at +4°C. Stable for one year. |
| 5F8 | |
| Monoclonal | |
| Rat | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title