Learn More
RFP Rat, Unconjugated, Clone: 5F8, Proteintech ChromoTek
Rat Monoclonal Antibody
Brand:  Chromotek 5F8-100
 View more versions of this product
Product Code. 17383263
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Rat monoclonal [5F8] to Red Fluorescent Proteins (RFP)
Specifications
Specifications
RFP
Monoclonal
Unconjugated
100 μL
Shipped at ambient temperature. Store at +4°C. Stable for one year.
Immunofluorescence, ELISA
5F8
Rat
Primary
IgG2a