missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFP Mouse, Unconjugated, Clone: 6G6, Proteintech ChromoTek
Mouse Monoclonal Antibody
£137.40 - £178.00
Specifications
| Antigen | RFP |
|---|---|
| Clone | 6G6 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
Mouse monoclonal [6G6] to RFP
Specifications
| RFP | |
| Western Blot | |
| Unconjugated | |
| IgG2c | |
| Shipped at ambient temperature. Store at +4°C. Stable for one year. |
| 6G6 | |
| Monoclonal | |
| Mouse | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title