Learn More
RFP Mouse, Unconjugated, Clone: 6G6, Proteintech ChromoTek
Mouse Monoclonal Antibody
Brand: Chromotek 6g6-100
This item is not returnable.
View return policy
Description
Mouse monoclonal [6G6] to RFP
Specifications
| RFP | |
| Monoclonal | |
| Unconjugated | |
| 100 μL | |
| Shipped at ambient temperature. Store at +4°C. Stable for one year. |
| Western Blot | |
| 6G6 | |
| Mouse | |
| Primary | |
| IgG2c |