missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ RF3000 Nose Cone

Product Code. 12934417 Shop All Fisherbrand Products
Change view
Click to view available options
:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12934417 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12934417

Please to purchase this item. Need a web account? Register with us today!

Stabilizes pipets while dispensing. Fisherbrand™ RF3000 Nose Cone and can be used with an adapter for smaller volumes and thinner pipets.

  • For use with RF3000™ Pipet Controller
  • Autoclavable
  • Available in Blue
  • Fits an adapter to stabilize smaller volumes and thinner pipets (separate purchase)

Specifications

Autoclavable Autoclavable
Color Assorted
For Use With RF3000, RF1000, Manual Controllers
Type Nose Cone
For Use With (Application) Pipetting
For Use With (Equipment) RF3000™ Pipet Controller
Quantity 5/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.