missing translation for 'onlineSavingsMsg'
Learn More

Rev-erb A alpha/NR1D1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£305.00 - £443.00

Specifications

Antigen Rev-erb A alpha/NR1D1
Dilution Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Frozen -Validated from a verified customer review.
Applications Immunocytochemistry, Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18420801
View Documents
Novus Biologicals
NBP1-84931-25ul
25 μL
£305.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18449440
View Documents
Novus Biologicals
NBP1-84931
0.1 mL
£443.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Rev-erb A alpha/NR1D1 Polyclonal antibody specifically detects Rev-erb A alpha/NR1D1 in Human, Amphibian samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen)
Specifications

Specifications

Rev-erb A alpha/NR1D1
Immunocytochemistry, Immunohistochemistry (Frozen)
Unconjugated
Rabbit
Human, Amphibian
EAR-1, EAR1nuclear receptor subfamily 1 group D member 1, HREV, nuclear receptor subfamily 1, group D, member 1, Rev-erbA-alpha, Rev-ErbAalpha, THRA1, THRALrev-erbA-alpha, V-erbA-related protein 1
This antibody was developed against Recombinant Protein corresponding to amino acids: TTLDSNNNTGGVITYIGSSGSSPSRTSPESLYSDNSNGSFQSLTQGCPTYFPPSPTGSLTQDPARSFGSIPPSLSDDGSPSSSSSSSSSSSSFYNGSPPGSLQVAMEDSSRVSPSKSTSNITKLNGMVLLCKVCGDVA
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Frozen -Validated from a verified customer review.
Polyclonal
Purified
RUO
PBS (pH 7.2) and 40% Glycerol
9572
IgG
Immunogen affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.