missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rev-erb A alpha/NR1D1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-84931
This item is not returnable.
View return policy
Description
Rev-erb A alpha/NR1D1 Polyclonal antibody specifically detects Rev-erb A alpha/NR1D1 in Human, Amphibian samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen)
Specifications
| Rev-erb A alpha/NR1D1 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Frozen -Validated from a verified customer review. | |
| EAR-1, EAR1nuclear receptor subfamily 1 group D member 1, HREV, nuclear receptor subfamily 1, group D, member 1, Rev-erbA-alpha, Rev-ErbAalpha, THRA1, THRALrev-erbA-alpha, V-erbA-related protein 1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: TTLDSNNNTGGVITYIGSSGSSPSRTSPESLYSDNSNGSFQSLTQGCPTYFPPSPTGSLTQDPARSFGSIPPSLSDDGSPSSSSSSSSSSSSFYNGSPPGSLQVAMEDSSRVSPSKSTSNITKLNGMVLLCKVCGDVA | |
| 0.1 mL | |
| Primary | |
| Human, Amphibian | |
| Purified |
| Immunocytochemistry, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 9572 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction