Learn More
Fisherbrand™ Reusable Dispenser Tips
Product Code. 11542363 Shop All Fisherbrand Products
Change view
Click to view available options
:
Capacity: 50mL
Product Code.
11542363 Capacity: 50mL
1 options
Product Code. 11542363
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Robust, stable, and durable dispenser tips for several brands of pipettors.
These tips are for use with the following pipettors:
  • Brand Handystep™ and Handystep™ electronic All tips
  • Eppendorf™ Multipette™ (not Multipette™ Plus) All tips
  • Gilson Distriman™
  • Ritter Ripette™ All tips
  • Tri-continent Easystep™
Additonal features:
  • These Dispenser™ tips are non-sterile and available by gamma irradiation
  • These dispenser tips are re-useable
  • The adapter for this dispenser tip is autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Includes With one autoclavable adapter
Quantity 25/Pk.
Volume (Metric) 50 mL
Type Dispenser Tip
For Use With (Equipment) Brand Handystep and Handystep, Electronic Eppendorf, Multipette (not Multipette Plus), Gilson Distriman, Ritter Ripette. Tri-continent Easystep